Assay Details
Target Gene Details
Entrez Gene ID: | 4774 |
Gene Name: | nuclear factor I A |
Gene Aliases: |
CTF, NF-I/A, NF1-A, NFI-A, NFI-L |
Location: |
Chr.1:61077124-61462788 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFIA | NM_001134673.3 | NP_001128145.1 | ||
| NM_001145511.1 | NP_001138983.1 | |||
| NM_001145512.1 | NP_001138984.1 | |||
| NM_005595.4 | NP_005586.1 | |||
| XM_011541512.2 | XP_011539814.1 | |||
| XM_011541514.2 | XP_011539816.1 | |||
| XM_011541515.2 | XP_011539817.1 | |||
| XM_017001362.1 | XP_016856851.1 | |||
| AB037860.1 | ||||
| AK299289.1 | ||||
| AK299356.1 | ||||
| AK299579.1 | ||||
| BC022264.1 | AAH22264.1 | |||
| U07809.1 | AAA93124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012657 | Chr.1:60299584 - 61316101 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map