Assay Details
Target Gene Details
Entrez Gene ID: | 51514 |
Gene Name: | denticleless E3 ubiquitin protein ligase homolog |
Gene Aliases: |
CDT2, DCAF2, L2DTL, RAMP |
Location: |
Chr.1:212035553-212105006 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTL | NM_001286229.1 | NP_001273158.1 | ||
| NM_001286230.1 | NP_001273159.1 | |||
| NM_016448.3 | NP_057532.3 | |||
| XM_011509614.1 | XP_011507916.1 | |||
| AF195765.1 | AAF35182.1 | |||
| AF345896.1 | AAK54706.1 | |||
| AK000742.1 | BAA91355.1 | |||
| AK027651.1 | BAB55267.1 | |||
| AK292343.1 | ||||
| AK299649.1 | ||||
| AK303346.1 | ||||
| BC033297.1 | AAH33297.1 | |||
| BC033540.1 | AAH33540.1 | |||
| DQ641253.1 |
More Information
Set Membership: |
|

Genomic Map