Assay Details
Target Gene Details
Entrez Gene ID: | 91734 |
Gene Name: | isopentenyl-diphosphate delta isomerase 2 |
Gene Aliases: |
IPPI2 |
Location: |
Chr.10:1018907-1025859 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI2 | NM_033261.2 | NP_150286.1 | ||
| AF271725.1 | AAK49437.1 | |||
| AK056950.1 | BAB71322.1 | |||
| BC017778.1 | AAH17778.1 |
Target Gene Details
Entrez Gene ID: | 55853 |
Gene Name: | IDI2 antisense RNA 1 |
Gene Aliases: |
C10orf110, HT009, IDI2-AS |
Location: |
Chr.10:1022637-1044201 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDI2-AS1 | NR_024628.1 | |||
| NR_024629.1 | ||||
| NR_027708.1 | ||||
| NR_027709.1 | ||||
| AF220183.1 | AAF67648.1 | |||
| AK314921.1 | ||||
| BF724542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv1046220 | Chr.10:1018084 - 1143058 on Build GRCh38 | Gain |
|
| esv3622095 | Chr.10:1018212 - 1031287 on Build GRCh38 | Loss |
|
| esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
| nsv549767 | Chr.10:1019000 - 1027744 on Build GRCh38 | Loss |
|
| nsv1161921 | Chr.10:823011 - 1033401 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map