Assay Details
Target Gene Details
Entrez Gene ID: | 10225 |
Gene Name: | CD96 molecule |
Gene Aliases: |
TACTILE |
Location: |
Chr.3:111542079-111665996 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD96 | NM_001318889.1 | NP_001305818.1 | ||
| NM_005816.4 | NP_005807.1 | |||
| NM_198196.2 | NP_937839.1 | |||
| NR_134917.1 | ||||
| XM_005247063.3 | XP_005247120.1 | |||
| XM_006713469.3 | XP_006713532.1 | |||
| XM_006713470.3 | XP_006713533.1 | |||
| XM_017005521.1 | XP_016861010.1 | |||
| XM_017005522.1 | XP_016861011.1 | |||
| AK290719.1 | ||||
| AK315237.1 | ||||
| AL833681.1 | CAI46155.1 | |||
| BC020749.1 | AAH20749.1 |
Target Gene Details
Entrez Gene ID: | 79413 |
Gene Name: | zinc finger BED-type containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:111592900-111595335 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBED2 | NM_024508.4 | 2 | 1564 | NP_078784.2 |
| BC003536.1 | 1 | 749 | AAH03536.1 | |
| BC071956.1 | 1 | 1029 | AAH71956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591275 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
|
| nsv997756 | Chr.3:111507211 - 111657738 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map