Assay Details
Target Gene Details
Entrez Gene ID: | 80324 |
Gene Name: | pseudouridylate synthase 1 |
Gene Aliases: |
MLASA1 |
Location: |
Chr.12:131929200-131944040 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PUS1 | NM_001002019.2 | 6 | 1413 | NP_001002019.1 |
| NM_001002020.2 | 6 | 1442 | NP_001002020.1 | |
| NM_025215.5 | 6 | 1790 | NP_079491.2 | |
| XM_011538768.2 | 5 | 1055 | XP_011537070.1 | |
| AF116238.1 | 5 | 1150 | AAD21042.1 | |
| AF318369.1 | 6 | 1782 | AAL55876.1 | |
| AI566545.1 | 1 | 225 | ||
| AI884644.1 | 1 | 225 | ||
| AK074659.1 | 6 | 1790 | ||
| AK292242.1 | 6 | 1386 | ||
| BC002901.2 | 6 | 1356 | AAH02901.1 | |
| BC009505.2 | 6 | 1561 | ||
| BC019320.1 | 6 | 1561 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560930 | Chr.12:131819514 - 131990296 on Build GRCh38 | Loss |
|
| esv3631282 | Chr.12:131915190 - 132065739 on Build GRCh38 | Gain |
|
| esv2751042 | Chr.12:131776139 - 132005401 on Build GRCh38 | Gain |
|
| nsv560928 | Chr.12:131793985 - 132060098 on Build GRCh38 | Gain |
|
| esv3631281 | Chr.12:131909102 - 131947534 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map