Assay Details
Target Gene Details
Entrez Gene ID: | 5368 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ |
Location: |
Chr.8:28316979-28343355 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNOC | NM_001284244.1 | 3 | 728 | NP_001271173.1 |
| NM_006228.4 | 4 | 921 | NP_006219.1 | |
| XM_005273532.2 | 4 | 950 | XP_005273589.1 | |
| XM_011544559.2 | 4 | 753 | XP_011542861.1 | |
| XM_017013578.1 | 3 | 1401 | XP_016869067.1 | |
| BC034758.1 | 4 | 766 | AAH34758.1 | |
| CA412176.1 | 1 | 295 | ||
| U48263.1 | 4 | 924 | AAC50651.1 | |
| X97370.1 | 3 | 736 | CAA66040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain |
|
| nsv819240 | Chr.8:28342879 - 28343407 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map