Assay Details
Target Gene Details
Entrez Gene ID: | 55107 |
Gene Name: | anoctamin 1 |
Gene Aliases: |
DOG1, ORAOV2, TAOS2, TMEM16A |
Location: |
Chr.11:70078302-70189545 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANO1 | NM_018043.5 | 1 | 333 | NP_060513.5 |
| XM_011545121.2 | 1 | 41 | XP_011543423.1 | |
| XM_011545123.2 | 1 | 41 | XP_011543425.1 | |
| XM_011545124.2 | 1 | 41 | XP_011543426.1 | |
| XM_011545125.2 | 1 | 41 | XP_011543427.1 | |
| XM_011545126.2 | 1 | 41 | XP_011543428.1 | |
| XM_011545127.2 | 1 | 41 | XP_011543429.1 | |
| XM_011545128.2 | 1 | 41 | XP_011543430.1 | |
| XM_017017956.1 | 1 | 41 | XP_016873445.1 | |
| XM_017017957.1 | 1 | 41 | XP_016873446.1 | |
| AB845669.1 | ||||
| AK293088.1 | 1 | 41 | ||
| AL833582.1 | 1 | 77 | ||
| AY728143.1 | 1 | 333 | AAU82085.1 | |
| CF593428.1 | 1 | 123 | ||
| KC577595.1 | 2 | 352 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1118217 | Chr.11:70078194 - 70078994 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map