Assay Details
Target Gene Details
Entrez Gene ID: | 153201 |
Gene Name: | solute carrier family 36 member 2 |
Gene Aliases: |
PAT2, TRAMD1 |
Location: |
Chr.5:151314978-151347590 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC36A2 | NM_181776.2 | 10 | 2318 | NP_861441.2 |
| XM_017009084.1 | 9 | 2226 | XP_016864573.1 | |
| AY162214.1 | 10 | 2286 | AAO11788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032558 | Chr.5:151257308 - 151325163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map