Assay Details
Target Gene Details
Entrez Gene ID: | 6609 |
Gene Name: | sphingomyelin phosphodiesterase 1 |
Gene Aliases: |
ASM, ASMASE, NPD |
Location: |
Chr.11:6390301-6394998 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPD1 | NM_000543.4 | NP_000534.3 | ||
| NM_001007593.2 | NP_001007594.2 | |||
| NM_001318087.1 | NP_001305016.1 | |||
| NM_001318088.1 | NP_001305017.1 | |||
| NR_027400.2 | ||||
| NR_134502.1 | ||||
| XM_011520303.1 | XP_011518605.1 | |||
| AA746054.1 | ||||
| AB209775.1 | BAD93012.1 | |||
| BC041164.1 | AAH41164.1 | |||
| BM977655.1 | ||||
| M59916.1 | AAA58377.1 | |||
| X52678.1 | CAA36901.1 | |||
| X52679.1 | CAA36902.1 | |||
| X59960.1 | CAA42584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553371 | Chr.11:6374460 - 6413235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map