Assay Details
Target Gene Details
Entrez Gene ID: | 124583 |
Gene Name: | calcium activated nucleotidase 1 |
Gene Aliases: |
DBQD, DBQD1, SCAN-1, SCAN1, SHAPY |
Location: |
Chr.17:78991716-79009817 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CANT1 | NM_001159772.1 | 6 | 1536 | NP_001153244.1 |
| NM_001159773.1 | 5 | 1397 | NP_001153245.1 | |
| NM_138793.3 | 4 | 1592 | NP_620148.1 | |
| XM_005257021.1 | 5 | 1660 | XP_005257078.1 | |
| XM_005257022.1 | 6 | 1515 | XP_005257079.1 | |
| XM_006721683.1 | 7 | 1683 | XP_006721746.1 | |
| XM_011524291.1 | 7 | 1988 | XP_011522593.1 | |
| XM_011524293.1 | 6 | 1878 | XP_011522595.1 | |
| XM_011524294.1 | 5 | 1353 | XP_011522596.1 | |
| XM_011524295.2 | 7 | 1651 | XP_011522597.1 | |
| AB097006.1 | BAC77359.1 | |||
| AB097033.1 | BAC77386.1 | |||
| AF328554.2 | 6 | 1498 | AAM94564.1 | |
| AJ312208.1 | 3 | 677 | CAC85468.1 | |
| AK074687.1 | 4 | 1531 | BAC11139.1 | |
| AK295930.1 | 6 | 1220 | ||
| BC005104.1 | AAH05104.1 | |||
| BC017655.1 | 5 | 1310 | AAH17655.1 | |
| BC065038.1 | 4 | 1213 | AAH65038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952378 | Chr.17:78989919 - 79007518 on Build GRCh38 | Deletion |
|
| nsv470610 | Chr.17:78973800 - 79091723 on Build GRCh38 | Loss |
|
| nsv523650 | Chr.17:78933797 - 79165563 on Build GRCh38 | Loss |
|
| nsv1057193 | Chr.17:78921031 - 79481744 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map