Assay Details
Target Gene Details
Entrez Gene ID: | 9397 |
Gene Name: | N-myristoyltransferase 2 |
Gene Aliases: |
- |
Location: |
Chr.10:15104588-15168732 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMT2 | NM_001308295.1 | 13 | 2290 | NP_001295224.1 |
| NM_004808.2 | 12 | 2233 | NP_004799.1 | |
| XM_005252642.4 | XP_005252699.1 | |||
| XM_006717539.3 | 11 | 2070 | XP_006717602.1 | |
| XM_017016947.1 | XP_016872436.1 | |||
| XM_017016948.1 | XP_016872437.1 | |||
| XM_017016949.1 | XP_016872438.1 | |||
| XM_017016950.1 | XP_016872439.1 | |||
| XM_017016951.1 | 12 | 2037 | XP_016872440.1 | |
| XM_017016952.1 | XP_016872441.1 | |||
| XM_017016953.1 | 13 | 2429 | XP_016872442.1 | |
| AF043325.1 | 12 | 2195 | AAC09295.1 | |
| AK094904.1 | 13 | 2208 | ||
| BC005232.1 | AAH05232.1 | |||
| BC006376.2 | 12 | 2151 | AAH06376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831794 | Chr.10:14993658 - 15148891 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map