Assay Details
Target Gene Details
Entrez Gene ID: | 170487 |
Gene Name: | actin like 10 |
Gene Aliases: |
C20orf134, dJ63M2.2 |
Location: |
Chr.20:33666498-33668525 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTL10 | NM_001024675.1 | 1 | 161 | NP_001019846.1 |
| BC137111.1 | 1 | 141 | ||
| BC137114.1 | 1 | 141 | ||
| BI832107.1 | 1 | 172 |
Target Gene Details
Entrez Gene ID: | 63941 |
Gene Name: | N-terminal EF-hand calcium binding protein 3 |
Gene Aliases: |
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5 |
Location: |
Chr.20:33657087-33674458 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAB3 | NM_031231.3 | NP_112508.3 | ||
| NM_031232.3 | NP_112509.3 | |||
| XM_005260510.1 | XP_005260567.1 | |||
| XM_011528992.2 | XP_011527294.1 | |||
| XM_017028015.1 | XP_016883504.1 | |||
| AB039947.1 | BAB16413.1 | |||
| AF193759.1 | AAG28415.1 | |||
| AF409141.1 | AAL01118.1 | |||
| AK023706.1 | BAB14649.1 | |||
| AK291895.1 | ||||
| AK309285.1 | ||||
| AL519842.3 | ||||
| BC047673.1 | AAH47673.1 | |||
| BQ212085.1 | ||||
| BQ431858.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954024 | Chr.20:33638795 - 33698694 on Build GRCh38 | Deletion |
|
| nsv1065189 | Chr.20:33597378 - 33807579 on Build GRCh38 | Loss |
|
| nsv833961 | Chr.20:33605380 - 33836992 on Build GRCh38 | Loss |
|
| nsv482067 | Chr.20:33666498 - 33668524 on Build GRCh38 | Gain |
|
| nsv833960 | Chr.20:33569059 - 33688060 on Build GRCh38 | Loss |
|
| dgv7560n54 | Chr.20:33657740 - 33667228 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map