Assay Details
Target Gene Details
Entrez Gene ID: | 10849 |
Gene Name: | CD3e molecule associated protein |
Gene Aliases: |
ASE-1, ASE1, CAST, PAF49 |
Location: |
Chr.19:45406129-45410766 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD3EAP | NM_001297590.1 | 1 | 119 | NP_001284519.1 |
| NM_012099.1 | 1 | 119 | NP_036231.1 | |
| XM_017026180.1 | 1 | 199 | XP_016881669.1 | |
| XM_017026181.1 | 1 | 199 | XP_016881670.1 | |
| BC054044.1 | AAH54044.1 | |||
| BU166593.1 | ||||
| U86751.1 | 1 | 119 | AAB68608.1 |
Target Gene Details
Entrez Gene ID: | 10848 |
Gene Name: | protein phosphatase 1 regulatory subunit 13 like |
Gene Aliases: |
IASPP, NKIP1, RAI, RAI4 |
Location: |
Chr.19:45379634-45406349 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R13L | NM_001142502.1 | NP_001135974.1 | ||
| XM_017026177.1 | XP_016881666.1 | |||
| XM_017026178.1 | XP_016881667.1 | |||
| XM_017026179.1 | XP_016881668.1 | |||
| BC064913.1 | AAH64913.1 | |||
| DB209153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| nsv833846 | Chr.19:45381005 - 45469089 on Build GRCh38 | Loss |
|
| dgv1054e212 | Chr.19:45347119 - 45407663 on Build GRCh38 | Gain |
|
| nsv579867 | Chr.19:45323108 - 45408744 on Build GRCh38 | Gain |
|
| nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map