Assay Details
Target Gene Details
Entrez Gene ID: | 11238 |
Gene Name: | carbonic anhydrase 5B |
Gene Aliases: |
CA-VB |
Location: |
Chr.X:15738281-15787625 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA5B | NM_007220.3 | 8 | 4990 | NP_009151.1 |
| AK057568.1 |
Target Gene Details
Entrez Gene ID: | 8551 |
Gene Name: | inactivation escape 2 (non-protein coding) |
Gene Aliases: |
NCRNA00011 |
Location: |
Chr.X:15785716-15787589 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INE2 | NR_002725.2 | 1 | 1007 | |
| Y10697.1 | 1 | 1007 |
More Information
Set Membership: |
|

Genomic Map