Assay Details
Target Gene Details
Entrez Gene ID: | 6297 |
Gene Name: | spalt like transcription factor 2 |
Gene Aliases: |
COLB, HSAL2, Sal-2, ZNF795, p150(Sal2) |
Location: |
Chr.14:21521080-21537216 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SALL2 | NM_001291446.1 | 4 | 3717 | NP_001278375.1 |
| NM_001291447.1 | 4 | 3697 | NP_001278376.1 | |
| NM_005407.2 | 2 | 4710 | NP_005398.2 | |
| NR_111966.1 | 3 | 1618 | ||
| NR_111967.1 | 3 | 1598 | ||
| XM_011537064.1 | 3 | 4048 | XP_011535366.1 | |
| XM_011537065.1 | 3 | 4194 | XP_011535367.1 | |
| AB002358.2 | 2 | 4697 | ||
| AK001276.1 | 3 | 1336 | ||
| BC024245.2 | 3 | 1411 | AAH24245.1 | |
| BC090958.1 | 3 | 1418 | AAH90958.1 | |
| BM698506.1 | 1 | 265 | ||
| X98834.1 | 2 | 4597 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633718 | Chr.14:21495207 - 21527525 on Build GRCh38 | Gain |
|
| esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map