Assay Details
Target Gene Details
Entrez Gene ID: | 1974 |
Gene Name: | eukaryotic translation initiation factor 4A2 |
Gene Aliases: |
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II |
Location: |
Chr.3:186783572-186789897 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4A2 | NM_001967.3 | NP_001958.2 | ||
| AF208852.1 | AAF64266.1 | |||
| AK130087.1 | ||||
| AK225646.1 | ||||
| AL117412.1 | CAI46218.1 | |||
| AL556981.3 | ||||
| BC012547.1 | AAH12547.1 | |||
| BC013708.1 | AAH13708.1 | |||
| BC015842.1 | AAH15842.1 | |||
| BC048105.1 | AAH48105.1 | |||
| BT009860.1 | AAP88862.1 | |||
| D30655.1 | BAA06336.1 | |||
| S79942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759206 | Chr.3:186619345 - 186849537 on Build GRCh38 | Loss |
|
| dgv5005n100 | Chr.3:186709551 - 186836503 on Build GRCh38 | Gain |
|
| nsv1012615 | Chr.3:186676249 - 186950429 on Build GRCh38 | Gain |
|
| esv34136 | Chr.3:186697095 - 187036942 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map