Assay Details
Target Gene Details
Entrez Gene ID: | 22838 |
Gene Name: | ring finger protein 44 |
Gene Aliases: |
- |
Location: |
Chr.5:176526699-176543456 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF44 | NM_014901.4 | 11 | 1861 | NP_055716.1 |
| XM_005265840.2 | 11 | 1573 | XP_005265897.1 | |
| XM_005265841.4 | 11 | 1818 | XP_005265898.1 | |
| XM_005265842.4 | 11 | 1803 | XP_005265899.1 | |
| XM_005265843.2 | 11 | 1509 | XP_005265900.1 | |
| XM_005265844.3 | 11 | 1614 | XP_005265901.1 | |
| XM_005265845.4 | 11 | 1752 | XP_005265902.1 | |
| XM_006714831.2 | 11 | 1564 | XP_006714894.1 | |
| XM_006714832.2 | 11 | 1432 | XP_006714895.1 | |
| XM_011534466.2 | 11 | 1576 | XP_011532768.1 | |
| XM_011534467.2 | 11 | 1807 | XP_011532769.1 | |
| AB029023.1 | 11 | 1746 | ||
| AK127673.1 | 2 | 683 | BAC87078.1 | |
| AK302385.1 | 11 | 1614 | ||
| AL547765.3 | 1 | 102 | ||
| BC039833.1 | 11 | 1817 | AAH39833.1 | |
| BC063297.1 | 11 | 1862 | AAH63297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516244 | Chr.5:176520050 - 176537482 on Build GRCh38 | Loss |
|
| nsv950722 | Chr.5:176517300 - 176533099 on Build GRCh38 | Deletion |
|
| nsv600344 | Chr.5:176527707 - 176544203 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map