Assay Details
Target Gene Details
Entrez Gene ID: | 10021 |
Gene Name: | hyperpolarization activated cyclic nucleotide gated potassium channel 4 |
Gene Aliases: |
SSS2 |
Location: |
Chr.15:73319859-73369264 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCN4 | NM_005477.2 | NP_005468.1 | ||
| XM_011521148.2 | XP_011519450.1 | |||
| AJ132429.1 | CAB42604.1 | |||
| AJ238850.1 | CAB52754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524049 | Chr.15:73320492 - 73387960 on Build GRCh38 | Loss |
|
| nsv569949 | Chr.15:73320036 - 73387960 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map