Assay Details
Target Gene Details
Entrez Gene ID: | 2304 |
Gene Name: | forkhead box E1 |
Gene Aliases: |
FKHL15, FOXE2, HFKH4, HFKL5, NMTC4, TITF2, TTF-2, TTF2 |
Location: |
Chr.9:97853255-97856715 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXE1 | NM_004473.3 | 1 | 2251 | NP_004464.2 |
| U89995.1 | 1 | 2279 | AAC51294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950914 | Chr.9:97850419 - 97857618 on Build GRCh38 | Deletion |
|
| nsv825000 | Chr.9:97850839 - 97856037 on Build GRCh38 | Gain |
|
| nsv6627 | Chr.9:97813976 - 97858752 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map