Assay Details
Target Gene Details
Entrez Gene ID: | 952 |
Gene Name: | CD38 molecule |
Gene Aliases: |
ADPRC 1, ADPRC1 |
Location: |
Chr.4:15778265-15853243 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD38 | NM_001775.3 | NP_001766.2 | ||
| NR_132660.1 | ||||
| AK297592.1 | ||||
| AK303169.1 | ||||
| AK313268.1 | ||||
| BC007964.2 | AAH07964.1 | |||
| CD688069.1 | ||||
| D84276.1 | BAA18964.1 | |||
| D84277.1 | BAA18965.1 | |||
| M34461.1 | AAA68482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893885 | Chr.4:15777609 - 15887721 on Build GRCh38 | Loss |
|
| dgv5145n100 | Chr.4:15761347 - 15834614 on Build GRCh38 | Gain |
|
| nsv10460 | Chr.4:15755988 - 15816583 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map