Assay Details
Target Gene Details
Entrez Gene ID: | 152404 |
Gene Name: | immunoglobulin superfamily member 11 |
Gene Aliases: |
BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3 |
Location: |
Chr.3:118900630-119146068 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF11 | NM_001015887.1 | 7 | 2816 | NP_001015887.1 |
| NM_152538.2 | 9 | 2947 | NP_689751.2 | |
| XM_006713517.1 | 8 | 2867 | XP_006713580.1 | |
| XM_011512462.2 | 8 | 3025 | XP_011510764.1 | |
| XM_011512463.1 | 9 | 2770 | XP_011510765.1 | |
| XM_011512464.2 | 9 | 2758 | XP_011510766.1 | |
| XM_011512465.1 | 9 | 2812 | XP_011510767.1 | |
| XM_011512466.2 | 8 | 2957 | XP_011510768.1 | |
| XM_011512467.2 | 8 | 2944 | XP_011510769.1 | |
| XM_011512468.1 | 10 | 2960 | XP_011510770.1 | |
| XM_011512469.1 | 8 | 2620 | XP_011510771.1 | |
| XM_011512470.1 | 8 | 2835 | XP_011510772.1 | |
| XM_011512471.2 | 8 | 2873 | XP_011510773.1 | |
| XM_011512472.1 | 8 | 2636 | XP_011510774.1 | |
| XM_017005789.1 | 7 | 2605 | XP_016861278.1 | |
| XM_017005790.1 | 8 | 2795 | XP_016861279.1 | |
| AB071618.1 | 7 | 2806 | BAD90695.1 | |
| AK122611.1 | 7 | 2707 | ||
| AL834256.1 | 5 | 2247 | CAD38931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011135 | Chr.3:116380194 - 119036472 on Build GRCh38 | Gain |
|
| nsv3960 | Chr.3:118883470 - 118914525 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map