Assay Details
Target Gene Details
Entrez Gene ID: | 9283 |
Gene Name: | G protein-coupled receptor 37 like 1 |
Gene Aliases: |
ET(B)R-LP-2, ETBR-LP-2 |
Location: |
Chr.1:202118669-202133588 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR37L1 | NM_004767.3 | 2 | 1454 | NP_004758.3 |
| XM_011510158.2 | 2 | 970 | XP_011508460.1 | |
| AK222639.1 | 2 | 1436 | BAD96359.1 | |
| AK295576.1 | 4 | 1437 | ||
| AK313044.1 | ||||
| BC050334.1 | 2 | 1454 | AAH50334.1 | |
| Y16280.1 | 2 | 1442 | CAA76153.1 |
More Information
Set Membership: |
|

Genomic Map