Assay Details
Target Gene Details
Entrez Gene ID: | 83694 |
Gene Name: | ribosomal protein S6 kinase like 1 |
Gene Aliases: |
RSKL2 |
Location: |
Chr.14:74903954-74923396 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KL1 | NM_031464.4 | 1 | 132 | NP_113652.2 |
| XM_011537206.2 | 2 | 392 | XP_011535508.1 | |
| XM_011537211.2 | 2 | 392 | XP_011535513.1 | |
| XM_011537213.2 | 2 | 392 | XP_011535515.1 | |
| XM_017021682.1 | XP_016877171.1 | |||
| XM_017021691.1 | 2 | 392 | XP_016877180.1 | |
| AK054800.1 | 2 | 251 | BAB70808.1 | |
| AK226137.1 |
More Information
Set Membership: |
|

Genomic Map