Assay Details
Target Gene Details
Entrez Gene ID: | 57646 |
Gene Name: | ubiquitin specific peptidase 28 |
Gene Aliases: |
- |
Location: |
Chr.11:113797875-113875580 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832273 | Chr.11:113784031 - 113957584 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142451643] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map