Assay Details
Target Gene Details
Entrez Gene ID: | 50 |
Gene Name: | aconitase 2 |
Gene Aliases: |
ACONM, HEL-S-284, ICRD, OCA8, OPA9 |
Location: |
Chr.22:41469125-41528989 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACO2 | NM_001098.2 | NP_001089.1 | ||
| XM_017028812.1 | XP_016884301.1 | |||
| AA724140.1 | ||||
| AK293562.1 | ||||
| AK296260.1 | ||||
| AK297213.1 | ||||
| AK301321.1 | ||||
| AK302691.1 | ||||
| AK314845.1 | ||||
| BC014092.2 | AAH14092.1 | |||
| BC026196.2 | AAH26196.1 | |||
| CR456365.1 | CAG30251.1 | |||
| CR536568.1 | CAG38805.1 | |||
| GQ472196.1 | ||||
| U80040.1 | AAB38416.1 |
Target Gene Details
Entrez Gene ID: | 171568 |
Gene Name: | RNA polymerase III subunit H |
Gene Aliases: |
RPC22.9, RPC8 |
Location: |
Chr.22:41525799-41544606 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3H | NM_001018050.3 | 6 | 2787 | NP_001018060.1 |
| NM_001018052.3 | 5 | 2700 | NP_001018062.1 | |
| NM_001282884.1 | 7 | 2515 | NP_001269813.1 | |
| NM_001282885.1 | 7 | 2474 | NP_001269814.1 | |
| NM_138338.4 | 7 | 2481 | NP_612211.1 | |
| AB051452.1 | 6 | 2642 | ||
| AK000365.1 | 1 | 355 | ||
| AK026524.1 | 7 | 2461 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss |
|
| nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| esv3647799 | Chr.22:41511654 - 41543254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map