Assay Details
Target Gene Details
Entrez Gene ID: | 5892 |
Gene Name: | RAD51 paralog D |
Gene Aliases: |
BROVCA4, R51H3, RAD51L3, TRAD |
Location: |
Chr.17:35099792-35119869 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51D | NM_001142571.1 | 5 | 749 | NP_001136043.1 |
| NM_002878.3 | 5 | 689 | NP_002869.3 | |
| NM_133629.2 | NP_598332.1 | |||
| NR_037711.1 | 4 | 570 | ||
| NR_037712.1 | ||||
| AB013341.1 | 5 | 539 | BAA25914.1 | |
| AB016223.1 | 4 | 420 | BAA31747.1 | |
| AB016224.1 | BAA31748.1 | |||
| AB016225.1 | BAA31749.1 | |||
| AB018360.1 | BAA33779.1 | |||
| AB018361.1 | BAA33780.1 | |||
| AB018362.1 | BAA33781.1 | |||
| AB018363.1 | 5 | 515 | BAA33782.1 | |
| AF034956.1 | 4 | 440 | AAC39719.1 | |
| AK296241.1 | 5 | 693 | ||
| AK301249.1 | 5 | 755 | ||
| AK302450.1 | 5 | 583 | ||
| AL117459.1 | CAB55937.1 | |||
| BC002723.2 | 4 | 454 | AAH02723.1 | |
| BC014422.1 | 5 | 568 | AAH14422.1 | |
| BI916871.1 | 5 | 689 | ||
| BX647297.1 | 5 | 636 | ||
| Y15572.1 | 5 | 552 | CAA75681.1 |
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51L3-RFFL | NR_037714.1 | |||
| AK097811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map