Assay Details
Target Gene Details
Entrez Gene ID: | 81846 |
Gene Name: | SET binding factor 2 |
Gene Aliases: |
CMT4B2, DENND7B, MTMR13 |
Location: |
Chr.11:9778667-10294216 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 44 - Exon 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBF2 | NM_030962.3 | NP_112224.1 | ||
| XM_005253154.4 | XP_005253211.1 | |||
| XM_005253155.4 | XP_005253212.1 | |||
| XM_011520394.2 | XP_011518696.1 | |||
| XM_017018372.1 | XP_016873861.1 | |||
| XM_017018373.1 | XP_016873862.1 | |||
| XM_017018374.1 | XP_016873863.1 | |||
| XM_017018375.1 | XP_016873864.1 | |||
| AK026571.1 | ||||
| AK091362.1 | ||||
| AK123621.1 | ||||
| AY234241.1 | AAO62733.1 | |||
| BC011143.1 | AAH11143.1 | |||
| BC043389.1 | AAH43389.1 | |||
| BC053867.1 | AAH53867.1 | |||
| BC063656.1 | AAH63656.1 | |||
| BC101466.1 | AAI01467.1 | |||
| BX538184.1 | CAD98056.1 | |||
| BX647533.1 | ||||
| CR749312.1 | CAH18167.1 |
Target Gene Details
Entrez Gene ID: | 283104 |
Gene Name: | SBF2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:9758293-9811319 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SBF2-AS1 | NR_036485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744095 | Chr.11:9623376 - 10133785 on Build GRCh38 | Deletion |
|
| nsv1053369 | Chr.11:9626290 - 9854507 on Build GRCh38 | Gain |
|
| nsv553468 | Chr.11:9646281 - 9806324 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map