Assay Details
Target Gene Details
Entrez Gene ID: | 100532735 |
Gene Name: | INO80B-WBP1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:74455023-74460891 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80B-WBP1 | NR_037849.1 |
Target Gene Details
Entrez Gene ID: | 23559 |
Gene Name: | WW domain binding protein 1 |
Gene Aliases: |
WBP-1 |
Location: |
Chr.2:74458400-74460891 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WBP1 | NM_012477.3 | NP_036609.1 | ||
| AA635803.1 | ||||
| AF087888.1 | AAP97187.1 | |||
| AK094297.1 | ||||
| BC010012.2 | AAH10012.1 | |||
| BC064482.1 | AAH64482.1 | |||
| BC071626.1 | AAH71626.1 | |||
| BC071850.1 | AAH71850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map