Assay Details
Target Gene Details
Entrez Gene ID: | 91807 |
Gene Name: | myosin light chain kinase 3 |
Gene Aliases: |
MLCK, MLCK2, caMLCK |
Location: |
Chr.16:46702282-46763246 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK3 | NM_001308301.1 | NP_001295230.1 | ||
| NM_182493.2 | NP_872299.2 | |||
| XM_006721334.3 | XP_006721397.1 | |||
| AB451455.1 | ||||
| AJ247087.1 | CAC42766.1 | |||
| AK299443.1 | ||||
| BC109097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638541 | Chr.16:46727423 - 46897599 on Build GRCh38 | Gain |
|
| nsv526656 | Chr.16:46505480 - 46812751 on Build GRCh38 | Gain |
|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
| esv3571142 | Chr.16:46430577 - 46840772 on Build GRCh38 | Gain |
|
| esv3892855 | Chr.16:46671431 - 46750308 on Build GRCh38 | Loss |
|
| esv3638540 | Chr.16:46678001 - 46738780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map