Assay Details
Target Gene Details
Entrez Gene ID: | 101929130 |
Gene Name: | uncharacterized LOC101929130 |
Gene Aliases: |
- |
Location: |
Chr.3:187291272-187297933 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929130 | NR_135551.1 | |||
| BX648496.1 |
Target Gene Details
Entrez Gene ID: | 5648 |
Gene Name: | mannan binding lectin serine peptidase 1 |
Gene Aliases: |
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF |
Location: |
Chr.3:187216085-187292220 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASP1 | NM_001031849.2 | 1 | 285 | NP_001027019.1 |
| NM_001879.5 | 1 | 285 | NP_001870.3 | |
| NM_139125.3 | 1 | 285 | NP_624302.1 | |
| NR_033519.1 | 1 | 285 | ||
| XM_006713701.2 | 1 | 252 | XP_006713764.1 | |
| XM_011512989.2 | 1 | 258 | XP_011511291.1 | |
| XM_011512990.2 | 1 | 258 | XP_011511292.1 | |
| XM_011512991.2 | 1 | 253 | XP_011511293.1 | |
| XM_017006869.1 | 1 | 257 | XP_016862358.1 | |
| XM_017006870.1 | 1 | 483 | XP_016862359.1 | |
| XM_017006871.1 | 1 | 258 | XP_016862360.1 | |
| AK129893.1 | ||||
| AK291157.1 | 1 | 121 | ||
| AK300663.1 | ||||
| AK304334.1 | ||||
| AL134380.1 | 1 | 227 | ||
| BC039724.1 | 1 | 220 | AAH39724.1 | |
| BX647237.1 | CAI46037.1 | |||
| CR749615.1 | 1 | 187 | CAH18409.1 | |
| DC423174.1 | 1 | 285 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009271 | Chr.3:187246858 - 187480081 on Build GRCh38 | Gain |
|
| nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map