Assay Details
Target Gene Details
Entrez Gene ID: | 9826 |
Gene Name: | Rho guanine nucleotide exchange factor 11 |
Gene Aliases: |
GTRAP48, PDZ-RHOGEF |
Location: |
Chr.1:156934840-157046903 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890682 | Chr.1:156899922 - 156951428 on Build GRCh38 | Loss |
|
| nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss |
|
| nsv3099 | Chr.1:156941668 - 156962079 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map