Assay Details
Target Gene Details
Entrez Gene ID: | 339105 |
Gene Name: | protease, serine 53 |
Gene Aliases: |
POL3S, UNQ308 |
Location: |
Chr.16:31083424-31090035 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS53 | NM_001039503.2 | 8 | 1151 | NP_001034592.1 |
| XM_011545816.1 | 7 | 2518 | XP_011544118.1 | |
| XM_011545817.1 | 7 | 2518 | XP_011544119.1 | |
| XM_011545818.2 | 7 | 2518 | XP_011544120.1 | |
| XM_011545819.1 | 8 | 2377 | XP_011544121.1 | |
| XM_011545820.1 | 8 | 2377 | XP_011544122.1 | |
| AJ627035.1 | 8 | 1151 | CAF25304.1 | |
| AK131087.1 | 2 | 4021 | BAC85137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv993244 | Chr.16:31080660 - 31090244 on Build GRCh38 | Deletion |
|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| nsv833193 | Chr.16:30949475 - 31122283 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map