Assay Details
Target Gene Details
Entrez Gene ID: | 374977 |
Gene Name: | maestro heat like repeat family member 7 |
Gene Aliases: |
C1orf175, HEATR8 |
Location: |
Chr.1:54641740-54710267 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MROH7 | NM_001039464.3 | 3 | 499 | NP_001034553.2 |
| NM_001291332.1 | NP_001278261.1 | |||
| NR_026782.2 | 3 | 493 | ||
| NR_111931.1 | 3 | 499 | ||
| AK093272.1 | 3 | 438 | BAC04116.1 | |
| AK130047.1 | 2 | 988 | BAC85285.1 | |
| AK301894.1 | ||||
| AK302446.1 | 3 | 446 | ||
| AK302703.1 | ||||
| AL704849.1 | ||||
| BC126126.1 | 1 | 199 | ||
| BC143891.1 | 1 | 199 | ||
| DB040876.1 | 3 | 309 | ||
| DB457600.2 | 3 | 484 |
Target Gene Details
Entrez Gene ID: | 100527960 |
Gene Name: | MROH7-TTC4 readthrough (NMD candidate) |
Gene Aliases: |
HEATR8-TTC4 |
Location: |
Chr.1:54641740-54742657 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MROH7-TTC4 | NR_037639.2 | 3 | 604 | |
| NR_037640.2 | 3 | 499 | ||
| NR_037641.2 | 3 | 499 | ||
| BX648088.1 | 3 | 438 | ||
| CR749830.1 | 3 | 556 | CAH18687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004778 | Chr.1:54635180 - 54764639 on Build GRCh38 | Gain |
|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |
|
| nsv546269 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain |
|
| dgv39e212 | Chr.1:54647335 - 54655840 on Build GRCh38 | Loss |
|
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
| nsv910 | Chr.1:54605786 - 54663322 on Build GRCh38 | Deletion |
|
| nsv829904 | Chr.1:54548613 - 54721384 on Build GRCh38 | Loss |
|

Genomic Map