Assay Details
Target Gene Details
Entrez Gene ID: | 64116 |
Gene Name: | solute carrier family 39 member 8 |
Gene Aliases: |
BIGM103, CDG2N, LZT-Hs6, PP3105, ZIP8 |
Location: |
Chr.4:102251041-102345498 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A8 | NM_001135146.1 | 9 | 2594 | NP_001128618.1 |
| NM_001135147.1 | NP_001128619.1 | |||
| NM_001135148.1 | 8 | 2031 | NP_001128620.1 | |
| NM_022154.5 | 8 | 2716 | NP_071437.3 | |
| XM_005263177.1 | 9 | 2503 | XP_005263234.1 | |
| XM_011532182.1 | 7 | 2529 | XP_011530484.1 | |
| XM_017008540.1 | 9 | 2511 | XP_016864029.1 | |
| XM_017008541.1 | 11 | 2888 | XP_016864030.1 | |
| XM_017008542.1 | XP_016864031.1 | |||
| AB020969.1 | 1 | 362 | ||
| AB040120.1 | 8 | 2664 | BAB21559.1 | |
| AK027652.1 | 8 | 2031 | BAB55268.1 | |
| AK304274.1 | ||||
| AL049963.2 | 1 | 692 | ||
| BC001320.1 | 2 | 1098 | AAH01320.1 | |
| BC012125.1 | 9 | 2570 | AAH12125.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5348n100 | Chr.4:102253039 - 102285398 on Build GRCh38 | Gain |
|
| nsv428447 | Chr.4:102182573 - 102340950 on Build GRCh38 | Gain |
|
| dgv5346n100 | Chr.4:102232552 - 102285398 on Build GRCh38 | Gain |
|
| nsv998689 | Chr.4:102258631 - 102285398 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map