Assay Details
Target Gene Details
Entrez Gene ID: | 8720 |
Gene Name: | membrane bound transcription factor peptidase, site 1 |
Gene Aliases: |
PCSK8, S1P, SKI-1 |
Location: |
Chr.16:84053761-84116943 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBTPS1 | NM_003791.3 | 23 | 3869 | NP_003782.1 |
| XM_017023816.1 | 22 | 3618 | XP_016879305.1 | |
| XM_017023817.1 | 24 | 3872 | XP_016879306.1 | |
| AK054744.1 | 3 | 2835 | ||
| AK091212.1 | 12 | 2579 | ||
| AK291773.1 | 23 | 3752 | ||
| AK299067.1 | 22 | 3094 | ||
| AL133583.1 | 8 | 1134 | CAB63727.1 | |
| BC114555.1 | AAI14556.1 | |||
| BC114961.1 | AAI14962.1 | |||
| BM048055.1 | 4 | 656 | ||
| BM680126.1 | 1 | 525 | ||
| D42053.1 | 23 | 3832 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892924 | Chr.16:84014717 - 84091665 on Build GRCh38 | Gain |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain |
|
| nsv1909 | Chr.16:84031468 - 84076703 on Build GRCh38 | Deletion |
|
| dgv546e214 | Chr.16:84011032 - 84095132 on Build GRCh38 | Gain |
|
| nsv573421 | Chr.16:84013110 - 84091665 on Build GRCh38 | Gain |
|
| nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss |
|
| nsv952061 | Chr.16:84049196 - 84062895 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map