Assay Details
Target Gene Details
Entrez Gene ID: | 1737 |
Gene Name: | dihydrolipoamide S-acetyltransferase |
Gene Aliases: |
DLTA, PDC-E2, PDCE2 |
Location: |
Chr.11:112024814-112064278 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLAT | NM_001931.4 | 7 | 1682 | NP_001922.2 |
| XM_011542647.2 | 7 | 1329 | XP_011540949.1 | |
| AK057299.1 | 7 | 1682 | ||
| AK223596.1 | 7 | 1071 | BAD97316.1 | |
| AK296274.1 | 7 | 961 | ||
| AK297103.1 | ||||
| AK299562.1 | 5 | 519 | ||
| AK303930.1 | 8 | 907 | ||
| AL832703.1 | 7 | 1053 | ||
| BC039084.1 | 7 | 1051 | AAH39084.1 | |
| X13822.1 | CAA32052.1 | |||
| Y00978.1 | 7 | 927 | CAA68787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map