Assay Details
Target Gene Details
Entrez Gene ID: | 6782 |
Gene Name: | heat shock protein family A (Hsp70) member 13 |
Gene Aliases: |
STCH |
Location: |
Chr.21:14371115-14383188 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA13 | NM_006948.4 | 2 | 279 | NP_008879.3 |
| AK128540.1 | 2 | 237 | ||
| AK223282.1 | 2 | 237 | BAD97002.1 | |
| AK299004.1 | 2 | 228 | ||
| AK312396.1 | 2 | 234 | ||
| BC036370.2 | 2 | 241 | AAH36370.1 | |
| BI548162.1 | 2 | 307 | ||
| U04735.1 | 2 | 246 | AAA16954.1 |
More Information
Set Membership: |
|

Genomic Map