Assay Details
Target Gene Details
Entrez Gene ID: | 51700 |
Gene Name: | cytochrome b5 reductase 2 |
Gene Aliases: |
B5R.2 |
Location: |
Chr.11:7665095-7678568 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5R2 | NM_001302826.1 | 9 | 1482 | NP_001289755.1 |
| NM_001302827.1 | 8 | 952 | NP_001289756.1 | |
| NM_016229.4 | 9 | 1136 | NP_057313.2 | |
| NR_126508.1 | 10 | 1249 | ||
| XM_005252975.4 | 9 | 1088 | XP_005253032.1 | |
| XM_006718251.2 | 9 | 1113 | XP_006718314.1 | |
| XM_011520184.2 | 9 | 1303 | XP_011518486.1 | |
| XM_011520185.2 | 12 | 1579 | XP_011518487.1 | |
| XM_017017920.1 | 11 | 1396 | XP_016873409.1 | |
| XM_017017921.1 | 10 | 1298 | XP_016873410.1 | |
| XM_017017922.1 | 8 | 930 | XP_016873411.1 | |
| AA814725.1 | 1 | 211 | ||
| AB209000.1 | 3 | 7253 | BAD92237.1 | |
| AF169802.1 | 9 | 1039 | AAF04811.1 | |
| AI342011.1 | 1 | 222 | ||
| AK290102.1 | 9 | 1455 | ||
| AL133582.1 | 8 | 872 | CAB63726.1 | |
| BF061336.1 | 1 | 232 | ||
| BM968347.1 | 1 | 229 | ||
| HY211860.1 | 1 | 206 |
Target Gene Details
Entrez Gene ID: | 8495 |
Gene Name: | PPFIA binding protein 2 |
Gene Aliases: |
Cclp1 |
Location: |
Chr.11:7513765-7667329 on Build GRCh38 |
Assay Gene Location: | Within Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPFIBP2 | XM_011520415.2 | XP_011518717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625293 | Chr.11:7515685 - 7685956 on Build GRCh38 | Gain |
|
| esv3891925 | Chr.11:7596802 - 7700323 on Build GRCh38 | Gain |
|
| esv3625296 | Chr.11:7635550 - 7688660 on Build GRCh38 | Gain |
|
| esv29577 | Chr.11:7664688 - 7665678 on Build GRCh38 | Loss |
|
| nsv1035575 | Chr.11:7648656 - 7744807 on Build GRCh38 | Gain |
|
| dgv58n21 | Chr.11:7652157 - 7688219 on Build GRCh38 | Loss |
|
| nsv553407 | Chr.11:7664234 - 7665353 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76915129] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map