Assay Details
Target Gene Details
Entrez Gene ID: | 9531 |
Gene Name: | BCL2 associated athanogene 3 |
Gene Aliases: |
BAG-3, BIS, CAIR-1, MFM6 |
Location: |
Chr.10:119651347-119677819 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAG3 | NM_004281.3 | 1 | 398 | NP_004272.2 |
| XM_005270287.1 | 1 | 421 | XP_005270344.1 | |
| AF071218.2 | 1 | 398 | AAF69592.2 | |
| AF095193.2 | 1 | 398 | AAD16122.2 | |
| AF127139.1 | 1 | 370 | AAF26839.1 | |
| AK222800.1 | 1 | 389 | BAD96520.1 | |
| AK291333.1 | 1 | 156 | ||
| AK313026.1 | 1 | 343 | ||
| AL137582.1 | 1 | 218 | CAB70824.1 | |
| BC006418.1 | 1 | 376 | AAH06418.1 | |
| BC014656.1 | 1 | 103 | AAH14656.1 | |
| BC107786.1 | 1 | 202 | AAI07787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891891 | Chr.10:119615669 - 119657099 on Build GRCh38 | Gain |
|
| nsv825585 | Chr.10:119648080 - 119761531 on Build GRCh38 | Gain |
|
| nsv1045668 | Chr.10:119611032 - 119859464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map