Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD1C | NM_001318154.1 | NP_001305083.1 | ||
| NM_001322258.1 | NP_001309187.1 | |||
| XM_017015897.1 | XP_016871386.1 | |||
| XM_017015899.1 | XP_016871388.1 | |||
| XM_017015901.1 | XP_016871390.1 | |||
| XM_017015903.1 | XP_016871392.1 | |||
| BF515122.1 | ||||
| CB854120.1 |
Target Gene Details
Entrez Gene ID: | 221035 |
Gene Name: | receptor accessory protein 3 |
Gene Aliases: |
C10orf74, Yip2b |
Location: |
Chr.10:63521353-63625123 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REEP3 | NM_001001330.2 | NP_001001330.1 | ||
| XM_011539501.2 | XP_011537803.1 | |||
| XM_017015896.1 | XP_016871385.1 | |||
| AK289976.1 | ||||
| AY562241.1 | AAT70686.1 | |||
| BC010040.2 | AAH10040.1 | |||
| BC057832.1 | AAH57832.1 | |||
| BC068557.1 | AAH68557.1 | |||
| KJ534927.1 | ||||
| KJ535017.1 | ||||
| KJ535031.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551187 | Chr.10:63521221 - 63521923 on Build GRCh38 | Loss |
|
| nsv551184 | Chr.10:63501424 - 63719655 on Build GRCh38 | Gain |
|
| esv22964 | Chr.10:63521142 - 63522056 on Build GRCh38 | Loss |
|
| nsv1112591 | Chr.10:63521140 - 63521940 on Build GRCh38 | Deletion |
|
| dgv1202n54 | Chr.10:63521170 - 63521923 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map