Assay Details
Target Gene Details
Entrez Gene ID: | 29799 |
Gene Name: | yippee like 1 |
Gene Aliases: |
FKSG3 |
Location: |
Chr.22:21697536-21735834 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YPEL1 | NM_013313.4 | 5 | 911 | NP_037445.1 |
| NR_130910.1 | 5 | 1295 | ||
| AB098734.1 | 5 | 911 | BAD51375.1 | |
| AB098735.1 | 5 | 911 | BAD51376.1 | |
| AF060862.1 | 4 | 611 | AAC15461.1 | |
| AF305069.1 | 4 | 671 | AAL09353.1 | |
| AW006162.1 | 1 | 101 | ||
| AW161786.1 | 1 | 480 | ||
| BC034486.1 | 5 | 769 | AAH34486.1 | |
| BC074501.1 | 5 | 776 | AAH74501.1 | |
| BI559478.1 | 5 | 791 | ||
| CR456376.2 | CAG30262.1 | |||
| CR933630.1 | 4 | 5014 | CAI45937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv191511 | Chr.22:21698277 - 21704496 on Build GRCh38 | Deletion |
|
| nsv510804 | Chr.22:21670242 - 21778475 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map