Assay Details
Target Gene Details
Entrez Gene ID: | 9774 |
Gene Name: | BCL2 associated transcription factor 1 |
Gene Aliases: |
BTF, bK211L9.1 |
Location: |
Chr.6:136256863-136290645 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCLAF1 | NM_001077440.1 | 12 | 5335 | NP_001070908.1 |
| NM_001077441.1 | 13 | 4969 | NP_001070909.1 | |
| NM_001301038.1 | 13 | 5482 | NP_001287967.1 | |
| NM_014739.2 | 13 | 5488 | NP_055554.1 | |
| XM_005267237.3 | 12 | 5277 | XP_005267294.1 | |
| XM_017011529.1 | 13 | 4956 | XP_016867018.1 | |
| XM_017011530.1 | 12 | 5616 | XP_016867019.1 | |
| XM_017011531.1 | 12 | 4752 | XP_016867020.1 | |
| XM_017011532.1 | 14 | 5651 | XP_016867021.1 | |
| BX537576.1 | 1 | 4688 | ||
| BX648431.1 | 5 | 3150 | ||
| BX648878.1 | 9 | 4229 | ||
| D79986.1 | 13 | 5488 |
More Information
Set Membership: |
|

Genomic Map