Assay Details
Target Gene Details
Entrez Gene ID: | 3959 |
Gene Name: | galectin 3 binding protein |
Gene Aliases: |
90K, BTBD17B, CyCAP, M2BP, MAC-2-BP, TANGO10B, gp90 |
Location: |
Chr.17:78971253-78979979 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGALS3BP | NM_005567.3 | 6 | 1017 | NP_005558.1 |
| AK055977.1 | 5 | 680 | ||
| AK057776.1 | 5 | 648 | ||
| AK293183.1 | 5 | 691 | ||
| AK295442.1 | 5 | 445 | ||
| AK301040.1 | 5 | 925 | ||
| AK301444.1 | 5 | 665 | ||
| AK307665.1 | 6 | 938 | ||
| BC002403.2 | 6 | 950 | AAH02403.1 | |
| BC002998.1 | 6 | 950 | AAH02998.1 | |
| BC015761.1 | 6 | 982 | AAH15761.1 | |
| KU178164.1 | 5 | 838 | ||
| KU178165.1 | 4 | 585 | ||
| L13210.1 | 6 | 1017 | AAA36193.1 | |
| X79089.1 | 6 | 969 | CAA55699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576101 | Chr.17:78911271 - 78991681 on Build GRCh38 | Gain |
|
| nsv952377 | Chr.17:78923019 - 78984118 on Build GRCh38 | Deletion |
|
| nsv523650 | Chr.17:78933797 - 79165563 on Build GRCh38 | Loss |
|
| nsv1057193 | Chr.17:78921031 - 79481744 on Build GRCh38 | Gain |
|
| nsv2155 | Chr.17:78945535 - 78979944 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map