Assay Details
Target Gene Details
Entrez Gene ID: | 57150 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
C6orf162, dJ102H19.2 |
Location: |
Chr.6:87322588-87342328 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM8 | NM_001042493.2 | 4 | 383 | NP_001035958.1 |
| NM_001287445.1 | NP_001274374.1 | |||
| NM_020425.5 | 3 | 362 | NP_065158.3 | |
| AF311339.1 | 3 | 404 | AAK38513.1 | |
| AK311963.1 | ||||
| AL050201.1 | 2 | 329 | ||
| BC070260.1 | 3 | 344 | AAH70260.1 | |
| BG534658.1 | 3 | 363 | ||
| DA293899.1 | 4 | 354 |
More Information
Additional Information:
For this assay, SNP(s) [rs114629345] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map