Assay Details
Target Gene Details
Entrez Gene ID: | 9926 |
Gene Name: | lysophosphatidylglycerol acyltransferase 1 |
Gene Aliases: |
FAM34A, FAM34A1, NET8 |
Location: |
Chr.1:211743457-211830772 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPGAT1 | NM_001320808.1 | 8 | 2346 | NP_001307737.1 |
| NM_014873.2 | 8 | 1757 | NP_055688.1 | |
| XM_005273362.3 | 9 | 1905 | XP_005273419.1 | |
| XM_011510228.2 | 9 | 1794 | XP_011508530.1 | |
| XM_011510229.2 | 8 | 1677 | XP_011508531.1 | |
| XM_017003004.1 | 7 | 2909 | XP_016858493.1 | |
| D86960.1 | 8 | 1757 |
More Information
Set Membership: |
|

Genomic Map