Assay Details
Target Gene Details
Entrez Gene ID: | 78991 |
Gene Name: | prenylcysteine oxidase 1 like |
Gene Aliases: |
- |
Location: |
Chr.5:149358007-149375112 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCYOX1L | NM_001301054.1 | NP_001287983.1 | ||
| NM_001301057.1 | NP_001287986.1 | |||
| NM_024028.3 | NP_076933.3 | |||
| XM_011537680.1 | XP_011535982.1 | |||
| AF451985.1 | AAP97684.1 | |||
| AK075414.1 | BAC11604.1 | |||
| AK127281.1 | 1 | 629 | ||
| AL834220.1 | 1 | 454 | CAD38901.2 | |
| BC000014.2 | AAH00014.1 | |||
| DA368718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599948 | Chr.5:149346599 - 149413198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map