Assay Details
Target Gene Details
Entrez Gene ID: | 766 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
CAVII |
Location: |
Chr.16:66844379-66854149 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA7 | NM_001014435.1 | 7 | 1511 | NP_001014435.1 |
| NM_005182.2 | 7 | 1359 | NP_005173.1 | |
| XM_005256136.3 | 7 | 1344 | XP_005256193.1 | |
| XM_011523311.1 | 7 | 1349 | XP_011521613.1 | |
| XM_011523312.1 | 8 | 1432 | XP_011521614.1 | |
| AY075019.1 | 7 | 1366 | AAL78167.1 | |
| AY075020.1 | 7 | 1513 | AAL78168.1 | |
| BC033865.1 | 7 | 1315 | AAH33865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510690 | Chr.16:66817285 - 66858281 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map