Assay Details
Target Gene Details
Entrez Gene ID: | 81888 |
Gene Name: | hydroxypyruvate isomerase (putative) |
Gene Aliases: |
HT036 |
Location: |
Chr.1:43450607-43454241 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYI | NM_001190880.2 | NP_001177809.1 | ||
| NM_001243526.1 | NP_001230455.1 | |||
| NM_031207.5 | NP_112484.3 | |||
| XM_005271239.4 | XP_005271296.1 | |||
| XM_005271240.4 | XP_005271297.1 | |||
| XM_006710937.3 | XP_006711000.1 | |||
| XM_011542222.2 | XP_011540524.1 | |||
| AF284750.1 | AAG59852.1 | |||
| AF284751.1 | AAG59853.1 | |||
| AK096244.1 | ||||
| AY037165.1 | AAK67642.1 | |||
| AY775560.1 | AAV84474.1 | |||
| BC006140.1 | AAH06140.1 | |||
| BC019041.2 | AAH19041.1 | |||
| BM826556.1 | ||||
| CD672533.1 | ||||
| JF911821.1 |
Target Gene Details
Entrez Gene ID: | 23334 |
Gene Name: | seizure threshold 2 homolog (mouse) |
Gene Aliases: |
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B |
Location: |
Chr.1:43389884-43454247 on Build GRCh38 |
Assay Gene Location: | Within Exon 75 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SZT2 | NM_015284.3 | 71 | 11578 | NP_056099.3 |
| AB007936.2 | 57 | 9430 | BAA32312.2 | |
| AK126972.1 | 10 | 3480 | BAC86771.1 | |
| BC151232.1 | 57 | 9430 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438377 | Chr.1:43451738 - 43468018 on Build GRCh38 | Loss |
|
| nsv950701 | Chr.1:43418430 - 43453929 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map