Assay Details
Target Gene Details
Entrez Gene ID: | 27035 |
Gene Name: | NADPH oxidase 1 |
Gene Aliases: |
GP91-2, MOX1, NOH-1, NOH1 |
Location: |
Chr.X:100843324-100874345 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOX1 | NM_001271815.1 | 12 | 2094 | NP_001258744.1 |
| NM_007052.4 | 13 | 2205 | NP_008983.2 | |
| NM_013955.2 | 12 | 2058 | NP_039249.1 | |
| XM_017029407.1 | 11 | 1854 | XP_016884896.1 | |
| AF166326.1 | AAF23232.1 | |||
| AI285296.1 | 1 | 324 | ||
| AK292201.1 | 13 | 2160 |
More Information
Set Membership: |
|

Genomic Map