Assay Details
Target Gene Details
Entrez Gene ID: | 51386 |
Gene Name: | eukaryotic translation initiation factor 3 subunit L |
Gene Aliases: |
EIF3EIP, EIF3S11, EIF3S6IP, HSPC021, HSPC025, MSTP005 |
Location: |
Chr.22:37848774-37888782 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3L | NM_001242923.1 | NP_001229852.1 | ||
| NM_016091.3 | NP_057175.1 | |||
| XM_005261625.1 | XP_005261682.1 | |||
| XM_006724260.3 | XP_006724323.1 | |||
| XM_017028814.1 | XP_016884303.1 | |||
| AF077207.1 | AAD27002.1 | |||
| AF083243.1 | AAD39841.1 | |||
| AF109359.1 | AAQ13507.1 | |||
| AK056129.1 | ||||
| AK093540.1 | ||||
| AK129648.1 | ||||
| AK222496.1 | BAD96216.1 | |||
| AK222529.1 | BAD96249.1 | |||
| AK293629.1 | ||||
| AK298776.1 | ||||
| AK301323.1 | ||||
| AK302346.1 | ||||
| AK315533.1 | ||||
| AL357455.1 | ||||
| BC001101.2 | AAH01101.1 | |||
| BC007510.2 | AAH07510.1 | |||
| BC029265.1 | AAH29265.1 | |||
| CR456436.1 | CAG30322.1 | |||
| DA051005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
| esv3893482 | Chr.22:37832559 - 37927500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map